missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ EAR2 Polyclonal Antibody
GREENER_CHOICE

Artikelnummer. 16557791
Change view
Click to view available options
Menge:
100 μL
Packungsgröße:
100 Mikroliter
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge unitSize
16557791 100 μL 100 Mikroliter
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 16557791 Lieferant Invitrogen™ Lieferanten-Nr. PA590335

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Rabbit Polyclonal Antibody

Immunogen sequence: MAMVTGGWGG PGGDTNGVDK AGGYPRAAED DSASPPGAAS DAEPGDEERP GLQVD; Positive Samples: Mouse liver, Rat liver; Cellular Location: Nucleus.

Transcription factor predominantly involved in transcriptional repression. Binds to promoter/enhancer response elements that contain the imperfect 5'-AGGTCA-3' direct or inverted repeats with various spacings which are also recognized by other nuclear hormone receptors. Involved in modulation of hormonal responses. Represses transcriptional activity of the lutropin-choriogonadotropic hormone receptor/LHCGR gene, the renin/REN gene and the oxytocin-neurophysin/OXT gene. Represses the triiodothyronine-dependent and -independent transcriptional activity of the thyroid hormone receptor gene in a cell type-specific manner. The corepressing function towards thyroid hormone receptor beta/THRB involves at least in part the inhibition of THRB binding to triiodothyronine response elements (TREs) by NR2F6. Inhibits NFATC transcription factor DNA binding and subsequently its transcriptional activity. Acts as transcriptional repressor of IL-17 expression in Th-17 differentiated CD4(+) T cells and may be involved in induction and/or maintenance of peripheral immunological tolerance and autoimmunity. Involved in development of forebrain circadian clock; is required early in the development of the locus coeruleus (LC).
TRUSTED_SUSTAINABILITY

Spezifikation

Antigen EAR2
Anwendungen ELISA, Western Blot
Klassifikation Polyclonal
Konzentration 0.29 mg/mL
Konjugat Unconjugated
Zusammensetzung PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gen NR2F6
Gen-Zugriffsnummer O09017, P10588, P43136
Gen-Alias AV090102; avian erythroblastic leukemia viral (v-erb-a) oncogene homolog-like 2; COUP transcription factor 3; COUPg; COUP-TF3; EAR2; EAR-2; Erba related 2; ERBAL2; ERBA-related gene-2; Nr2f6; Nuclear receptor subfamily 2 group F member 6; nuclear receptor subfamily 2, group F, member 6; nuclear receptor V-erbA-related; orphan nuclear receptor EAR-2; Ovalbumin upstream promoter gamma nuclear receptor; ovalbumin upstream promoter gamma nuclear receptor rCOUPg; v-erb-a avian erythroblastic leukemia viral oncogene homolog-like 2; V-erbA-related protein 2; V-erbA-related protein EAR-2
Gensymbole NR2F6
Wirtsspezies Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-55 of human NR2F6 (NP_0052252).
Reinigungsverfahren Affinity chromatography
Menge 100 μL
Regulatorischer Status RUO
Primär oder sekundär Primary
Gen-ID (Entrez) 13864, 2063, 245980
Zielspezies Human, Mouse, Rat
Inhalt und Lagerung -20°C, Avoid Freeze/Thaw Cycles
Produkttyp Antibody
Form Liquid
Isotype IgG
Mehr anzeigen Weniger anzeigen
Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.