missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EDA Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 25892-1-AP-20UL
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
EDA is a type II membrane protein that can be cleaved by furin to produce a secreted form. This protein, which belongs to the tumor necrosis factor family, acts as a homotrimer and may be involved in cell-cell signaling during the development of ectodermal organs. Defects in the gene for EDA are a cause of ectodermal dysplasia, anhidrotic, which is also known as X-linked hypohidrotic ectodermal dysplasia.Spezifikation
EDA | |
Polyclonal | |
Unconjugated | |
EDA | |
Ectodermal dysplasia protein, ectodysplasin A, ED1, ED1 A1, ED1 A2, EDA, EDA protein, EDA1, EDA2, HED, XHED, XLHED | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
13607, 1896 | |
-20°C | |
Liquid |
Western Blot, Immunohistochemistry (Paraffin) | |
0.3 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O54693, Q92838 | |
EDA | |
EDA Fusion Protein Ag23154 | |
20 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |