missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human EDA2R/TNFRSF27/XEDAR Antibody, R&D Systems™
Goat Polyclonal Antibody has been used in 1 publication
N.V.
Spezifikation
Antigen | EDA2R/TNFRSF27/XEDAR |
---|---|
Konzentration | LYOPH |
Verdünnung | Western Blot 0.1 ug/mL, Blockade of Receptor-ligand Interaction |
Anwendungen | Western Blot, Blockade of Receptor-ligand Interaction |
Klassifikation | Polyclonal |
Beschreibung
EDA2R/TNFRSF27/XEDAR Polyclonal specifically detects EDA2R/TNFRSF27/XEDAR in Human samples. It is validated for Western Blot, Blockade of Receptor-ligand Interaction.Spezifikation
EDA2R/TNFRSF27/XEDAR | |
Western Blot 0.1 ug/mL, Blockade of Receptor-ligand Interaction | |
Polyclonal | |
Goat | |
Human | |
ectodysplasin A2 receptor, EDA-A2 receptor, EDAA2R, EDA-A2R, EDAA2Rtumor necrosis factor receptor superfamily member XEDAR, EDAR2, TNFRSF27, XEDAR, XEDARTNFRSF27tumor necrosis factor receptor superfamily member 27, X-linked ectodysplasin-A2 receptor | |
EDA2R | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Western Blot, Blockade of Receptor-ligand Interaction | |
Unconjugated | |
RUO | |
Q9HAV5 | |
60401 | |
Mouse myeloma cell line NS0-derived recombinant human EDA2R/TNFRSF27/XEDAR Met1-Glu136 Accession # Q9HAV5 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects human EDA2R/TNFRSF27/XEDAR in direct ELISAs and Western blots. |
For Research Use Only