Learn More
Ansell EDGE 48-700 Cut Protection Work Gloves
Product Code. 30259526 Shop All Ansell Products
Change view
Click to view available options
Size:
9
Unit Size:
12 pairs
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
30259526 9 12 pairs
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30259526 Supplier Ansell Supplier No. 48700090
Verfügbar ab: 03-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Medium-duty cut and abrasion-resistant gloves, offering high comfort and dexterity.
  • Enhanced dexterity: These comfortable high-dexterity gloves are designed with a seamless liner and knitted wrist to ensure a good level of flexibility and close fit, and may also be worn as an underglove
  • Improved durability: EDGE™ 48-700 cut protection work gloves feature a reinforced thumb crotch that increases abrasion resistance and wear life
  • Food handling compliance: In addition, these food gloves comply with FDA food handling requirements and the Commission Regulation (EC) No. 1935/2004 as well as the Regulation (EC) No. 2023/2006 related to Good Manufacturing Practices (GMP) for materials and articles intended to come in contact with foodstuffs
TRUSTED_SUSTAINABILITY
Specifications
Coating Material Uncoated
Color Gray
Cut Resistance Rating Level C
Hand Specific
Latex Free Yes
Material Polyethylene, Wrapped Yarn, Fiberglass, Spandex, Polyester
Product Type Industrial Gloves
Size 9