Learn More
EFR3A Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA554623
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: NTSSKDNDEK IVQNAIIQTI GFFGSNLPDY QRSEIMMFIM GKVPVFGTST HTLDISQLGD LGTRRIQIML L Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 96%.
EFR3A Polyclonal antibody specifically detects EFR3A in Human samples. It is validated for Immunohistochemistry.Spezifikation
EFR3A | |
Polyclonal | |
Unconjugated | |
EFR3A | |
A130089M23Rik; BB071175; C76891; C920006C10Rik; D030063F01Rik; EFR3 homolog A; EFR3 homolog A (S. cerevisiae); Efr3a; KIAA0143; mKIAA0143; Protein EFR3 homolog A; Protein EFR3-like; RGD1305976 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
23167 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q14156 | |
EFR3A | |
Recombinant protein corresponding to Human EFR3A | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only