missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EID1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA550401
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
The antibody detects endogenous levels of total EID1 protein.
EID1 Polyclonal antibody specifically detects EID1 in Human, Mouse samples. It is validated for Immunohistochemistry (Paraffin).Spezifikation
EID1 | |
Polyclonal | |
Unconjugated | |
EID1 | |
21 kDa pRb-associated protein; 2610002K20Rik; C15orf3; CREBBP/EP300 inhibitor 1; CREBBP/EP300 inhibitory protein 1; CRI1; E1A-like inhibitor of differentiation 1; EID; eid1; EID-1; EP300 interacting inhibitor of differentiation 1; EP300-interacting inhibitor of differentiation 1; IRO45620; NB4 apoptosis related protein; ORF12; PNAS-22; PTD014; Rb- and p300-binding protein EID-1; RBP21; retinoblastoma protein-associated protein; RGD1562702 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
23741, 58521 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.7 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
Q9DCR4, Q9Y6B2 | |
EID1 | |
Fusion protein corresponding to residues near the N terminal of human EP300 interacting inhibitor of differentiation 1 | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |