missing translation for 'onlineSavingsMsg'
Learn More

EIF3D Mouse anti-Human, Mouse, Rat, Clone: 2C5A3, Proteintech

Mouse Monoclonal Antibody

Marke:  Proteintech 66024-1-IG-150UL

Weitere Details : Netto-Gewicht : 0.00010kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 16887525

  • 329.85 € / 150 Mikroliter

Voraussichtliches Versanddatum: 18-06-2021

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Beschreibung

Beschreibung

eIF3D is a protein belonging to the largest of the eukaryotic translation initiation factor family, eukaryotic translation initiation factor-3 (eIF3). eIF3 is a multiprotein complex composed of at least 13 different subunits. eIF3D binds to the 40S ribosomal subunit and thus helps in maintaining the 40S and 60S ribosomal subunits in a dissociated state. An important role of eIF3D is the formation of the 40S initiation complex by interacting with the ternary complex of eIF2/GTP/methionyl-tRNA, and promotes mRNA binding. eIF3D is the major RNA binding subunit of the eIF3 complex. It associates with the subunit p170 of eIF-3. Ubiquitous expression of eIF3D is seen in most of the tissues and it is phosphorylated upon DNA damage, likely by ATM or ATR.
Spezifikation

Spezifikation

EIF3D
2C5A3
Unconjugated
EIF3D
Liquid
Eif3d
EIF3D Fusion Protein Ag18092
Protein A
RUO
Antibody
Monoclonal
Human, Mouse, Rat
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
0.78 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
O15371, O70194, Q6AYK8
eIF 3 zeta, eIF3 p66, eIF3 zeta, EIF3D, EIF3S7
Mouse
IgG2b
150 μL
-20°C
Primary
362952, 55944, 8664
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation