missing translation for 'onlineSavingsMsg'
Learn More

eIF3f Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Marke:  Thermo Scientific PA561918

Weitere Details : Netto-Gewicht : 0.02000kg

Artikelnummer. 15739721

  • 530.00 € / 100 Mikroliter
Verfügbar ab: 08-05-2024
Sie sich ein, um den Lagerbestand zu sehen

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Beschreibung

Beschreibung

Immunogen sequence: DSYERRNEGA ARVIGTLLGT VDKHSVEVTN CFSVPHNESE DEVAVDMEFA KNMYELHKKV SPNELILGWY A Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.

eIF3f Polyclonal antibody specifically detects eIF3f in Human samples. It is validated for Immunohistochemistry, Western Blot.
Spezifikation

Spezifikation

eIF3f
Polyclonal
Unconjugated
EIF3F
0610037M02Rik; AA409853; deubiquitinating enzyme eIF3f; eIF3 p47; eIF-3-epsilon; eIF3-epsilon; EIF3F; eIF3-p47; Eif3s5; eukaryotic translation initiation factor 3 subunit 5; eukaryotic translation initiation factor 3 subunit 5 epsilon; eukaryotic translation initiation factor 3 subunit F; eukaryotic translation initiation factor 3, subunit 5 (epsilon); eukaryotic translation initiation factor 3, subunit 5 (epsilon, 47kD); eukaryotic translation initiation factor 3, subunit 5 epsilon, 47kDa; eukaryotic translation initiation factor 3, subunit F
Rabbit
Antigen affinity chromatography
RUO
8665
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry, Western Blot
0.2 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
O00303
EIF3F
Recombinant protein corresponding to Human eIF3f
100 μL
Primary
Human
Antibody
IgG
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote

Research Use Only