missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ EIF4E (Human) Recombinant Protein
Human EIF4E full-length ORF ( AAH12611, 1 a.a. - 217 a.a.) recombinant protein with GST-tag at N-terminal.
Marke: Abnova™ H00001977-P01.25ug
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
- Sequence: MATVEPETTPTPNPPTTEEEKTESNQEVANPEHYIKHPLQNRWALWFFKNDKSKTWQANLRLISKFDTVEDFWALYNHIQLSSNLMPGCDYSLFKDGIEPMWEDEKNKRGGRWLITLNKQQRRSDLNRFWLETLLCLIGESFDDYSDDVCGAVVNVRAKGDKIAIWTTECENREAVTHIGRVYKERLGLPPKIVIGYQSHADTATKSGSTTKNRFVV
Spezifikation
AAH12611 | |
eukaryotic translation initiation factor 4E | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
EIF4E | |
GST |
1977 | |
Wheat germ expression system | |
25 μg | |
CBP, EIF4E1, EIF4EL1, EIF4F, MGC111573 | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |