missing translation for 'onlineSavingsMsg'
Learn More
Learn More
eIF5A Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA563944
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: MCGTGGTDSK TRRPPHRASF LKRLE Highest antigen sequence identity to the following orthologs: Mouse - 40%, Rat - 40%.
eIF5A Polyclonal antibody specifically detects eIF5A in Human samples. It is validated for Immunohistochemistry.Spezifikation
eIF5A | |
Polyclonal | |
Unconjugated | |
EIF5A | |
AA410058; D19Wsu54e; Eif4d; eIF-4D; Eif5a; eIF-5A; EIF5A1; eIF-5A1; eIF-5A-1; eIF5AI; eukaryotic initiation factor 5A; Eukaryotic initiation factor 5A isoform 1; eukaryotic translation initiation factor 5A; eukaryotic translation initiation factor 5A-1; MGC104255; MGC99547; OTTHUMP00000128384; Rev-binding factor; uORF A | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
1984 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P63241 | |
EIF5A | |
Recombinant protein corresponding to Human eIF5A | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only