Learn More
ACROBiosystems ELISA Assay Kit for Anti-PD-1 h-mAb in Mouse Serum
For quantification of monoclonal anti-PD-1 antibodies in mouse serum.
399.19 €
Spezifikation
Produkttyp | Pk. Assay Kit |
---|---|
Kitinhalt | The enclosed ELISA assay kit for anti-PD-1 h-mAb in mouse serum is based on an enzyme immunoassay (ELISA) between recombinant PD-1 protein (ECD) and a biotinylated anti-PD-1 monoclonal antibody. The method employs the principle of competitive ELISA, allow |
Versandbedingung | RT |
Lagerungsbedingungen | -20°C, aviod light |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
15806677
|
ACROBiosystems
EPM-V1-96TESTS |
96 Tests |
399.19 €
1 Set |
Verfügbar ab: 03-05-2024 Loggen Sie sich ein, um den Lagerbestand zu sehen |
Bitte melden Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute! | ||||
Beschreibung
How a therapeutic antibody is metabolized in the body is pertinently relevant to its efficacy. Therefore, pharmacokinetics study is an important part of the drug development. PD-1 is probably the most targeted molecule in today’s pharmaceutical industry, thanking to the clinical success of Keytruda® and Opdivo®. Many investigational anti-PD-1 mAbs are being developed. There is a growing need for a standard assay that can be used to facilitate the study of their pharmacokinetics.Spezifikation
Pk. Assay Kit | |
The enclosed ELISA assay kit for anti-PD-1 h-mAb in mouse serum is based on an enzyme immunoassay (ELISA) between recombinant PD-1 protein (ECD) and a biotinylated anti-PD-1 monoclonal antibody. The method employs the principle of competitive ELISA, allow | |
RT | |
-20°C, aviod light |