Learn More
ACROBiosystems ELISA Assay Kit for Anti-PD-1 h-mAb in Mouse Serum
For quantification of monoclonal anti-PD-1 antibodies in mouse serum.
399.19 €
Spezifikation
Produkttyp Pk. Assay Kit
Kitinhalt The enclosed ELISA assay kit for anti-PD-1 h-mAb in mouse serum is based on an enzyme immunoassay (ELISA) between recombinant PD-1 protein (ECD) and a biotinylated anti-PD-1 monoclonal antibody. The method employs the principle of competitive ELISA, allow
Versandbedingung RT
Lagerungsbedingungen -20°C, aviod light
Mehr Spezifikationen ansehen
Produkte 1
Produktcode Marke Menge Preis Menge & Verfügbarkeit  
Produktcode Marke Menge Preis Menge & Verfügbarkeit  
15806677
Unterlagen ansehen Aktionsdetails
ACROBiosystems
EPM-V1-96TESTS
96 Tests
399.19 €
1 Set
Alternative Produkte anzeigen
Sparen Sie bis zu 
»
Verfügbar ab: 03-05-2024
Sie sich ein, um den Lagerbestand zu sehen
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
Beschreibung
Beschreibung
How a therapeutic antibody is metabolized in the body is pertinently relevant to its efficacy. Therefore, pharmacokinetics study is an important part of the drug development. PD-1 is probably the most targeted molecule in today’s pharmaceutical industry, thanking to the clinical success of Keytruda® and Opdivo®. Many investigational anti-PD-1 mAbs are being developed. There is a growing need for a standard assay that can be used to facilitate the study of their pharmacokinetics.
Spezifikation
Spezifikation
Pk. Assay Kit
The enclosed ELISA assay kit for anti-PD-1 h-mAb in mouse serum is based on an enzyme immunoassay (ELISA) between recombinant PD-1 protein (ECD) and a biotinylated anti-PD-1 monoclonal antibody. The method employs the principle of competitive ELISA, allow
RT
-20°C, aviod light