Learn More
ELOA Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA552224
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: ERHLGEPHGK GVVSQNKEHK SSHKDKRPVD AKSDEKASVV SREKSHKALS KEENRRPPSG DNAREKPPSS GVKKEKDREG SSLKKKCLPP SEAASDNHLK KPKHRDPEKA KLDKSKQGLD SFDTGKGAGD Highest antigen sequence identity to the following orthologs: Mouse - 67%, Rat - 69%.
ELOA Polyclonal antibody specifically detects ELOA in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Spezifikation
ELOA | |
Polyclonal | |
Unconjugated | |
ELOA | |
110kDa; AA408125; El; EloA; EloA2; Elongin 110 kDa subunit; elongin A; Elongin A2; elongin-A; Elongin-A2; MSTP059; RNA polymerase II transcription factor elongin subunit A (110 kDa); RNA polymerase II transcription factor SIII subunit A; RNA polymerase II transcription factor SIII subunit A1; SIII; SIII p110; TCEB3; TCEB3A; TCEB3B; TCEB3L; transcription elongation factor B (SIII), polypeptide 3; transcription elongation factor B (SIII), polypeptide 3 (110kDa, elongin A); transcription elongation factor B alpha subunit; transcription elongation factor B polypeptide 3; transcription elongation factor B subunit 3 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
51224, 6924 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q14241, Q8IYF1 | |
ELOA, ELOA2 | |
Recombinant protein corresponding to Human ELOA | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only