Learn More
ELOA2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA560392
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: QKRPQHSHSN KKRPSLDGRD PGNGTHGLSP EEKEQLSNDR ETQEGKPPTA HLDRTSVSSL SEVEEVDMAE EFEQPTLSCE Highest antigen sequence identity to the following orthologs: Mouse - 36%, Rat - 44%.
This gene encodes the transcriptionally active subunit of the SIII (or elongin) transcription elongation factor complex, which also includes two regulatory subunits, elongins B and C. This complex acts to increase the rate of RNA chain elongation by RNA polymerase II by suppressing transient pausing of the polymerase at many sites along the DNA template. Whereas a related protein with similar function, elongin A, is ubiquitously expressed, the encoded protein is specifically expressed in the testis, suggesting it may have a role in spermatogenesis.Spezifikation
ELOA2 | |
Polyclonal | |
Unconjugated | |
ELOA2 | |
EloA2; Elongin-A2; HsT832; RNA polymerase II transcription factor SIII subunit A2; TCEB3B; TCEB3L; transcription elongation factor (SIII) elongin A2 elongin A2; transcription elongation factor B polypeptide 3B; transcription elongation factor B subunit 3B | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
51224 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.3 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8IYF1 | |
ELOA2 | |
Recombinant protein corresponding to Human ELOA2 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only