Beschreibung
ELSPBP1 Polyclonal specifically detects ELSPBP1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Spezifikation
Spezifikation
| Antigen | ELSPBP1 |
| Anwendungen | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Klassifikation | Polyclonal |
| Konjugat | DyLight 550 |
| Verdünnung | Immunohistochemistry, Immunohistochemistry-Paraffin |
| Zusammensetzung | 50mM Sodium Borate |
| Gen-Alias | E12epididymal protein 12, EDDM12, Epididymal secretory protein 12, epididymal sperm binding protein 1, epididymal sperm-binding protein 1, HE12 |
| Wirtsspezies | Rabbit |
| Immunogen | Produced in rabbits immunized with E. coli-derived Human ELSPBP1 fragment. |
| Reinigungsverfahren | Antigen Affinity-purified |
| Mehr anzeigen |