missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EMC6 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA564051
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: VVAKREGPPF ISEAAVRGNA AVLDYCRTS Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.
EMC6 Polyclonal antibody specifically detects EMC6 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin).Spezifikation
EMC6 | |
Polyclonal | |
Unconjugated | |
EMC6 | |
EMC6; ER membrane protein complex subunit 6; TMEM93; Transmembrane protein 93 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
83460 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin) | |
0.08 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9BV81 | |
EMC6 | |
Recombinant protein corresponding to Human EMC6 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only