Description
- This item is an accessory part for the Hydrosol Filter Holder, 47 mm, stainless steel.
Specifications
Specifications
| Autoclavable | Not Autoclavable |
| Disposable | Reusable |
| For Use With (Equipment) | Millipore 47 mm All-glass Vacuum Filter Holder |
| Material | PTFE |
| Sterility | Non-sterile |
| Type | Support Gasket |
| Diameter (Metric) | 47 mm |