Learn More
MilliporeSigma™ MQuant™ pH Indicator Papers
Measure pH onsite without instruments or extensive preparation
Marke: MilliporeSigma™ 1095570003
Weitere Details : Netto-Gewicht : 0.04200kg
Spezifikation
6.4 to 8 | |
6.4 to >8 | |
10°C to 25°C |
Environmental analysis and industrial in-process controls, calibration and checking of the products by certified buffer Solutions, rapid on-the-spot pH determination, non-bleeding even in strong alkaline solutions for accurate pH determination | |
3 x 4.8 Rolls | |
Special pH Indicator Paper |