Learn More
Endomucin Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA552229
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: EAANNSLVVT TTKPSITTPN TESLQKNVVT PTTGTTPKGT ITNELLKMSL MSTATFLTSK DEGLKATTTD VRKNDSIISN VTVTSVTLPN AVSTLQSSKP KTETQSSIKT TEIP Highest antigen sequence identity to the following orthologs: Mouse - 37%, Rat - 39%.
Endomucin Polyclonal antibody specifically detects Endomucin in Human samples. It is validated for Immunohistochemistry.Spezifikation
Endomucin | |
Polyclonal | |
Unconjugated | |
Emcn | |
0610012K22Rik; AI315669; Emcn; EMCN2; Endomucin; endomucin-1; endomucin-1/2; Endomucin-2; gastric cancer antigen Ga34; Muc14; MUC-14; mucin 14, endothelial; mucin-14; sialomucin | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
51705 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9ULC0 | |
Emcn | |
Recombinant protein corresponding to Human Endomucin | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only