missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ENPP4 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA550655
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
The antibody detects endogenous levels of total ENPP4 protein.
ENPP4 Polyclonal antibody specifically detects ENPP4 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Spezifikation
ENPP4 | |
Polyclonal | |
Unconjugated | |
ENPP4 | |
4933413N07Rik; AA986363; AI195364; AP3A hydrolase; AP3Aase; Bis(5'-adenosyl)-triphosphatase enpp4; ectonucleotide pyrophosphatase/phosphodiesterase 4; ectonucleotide pyrophosphatase/phosphodiesterase 4 (putative function); ectonucleotide pyrophosphatase/phosphodiesterase 4 (putative); ectonucleotide pyrophosphatase/phosphodiesterase family member 4; E-NPP 4; ENPP4; E-NPP4; Gm90; KIAA0879; mKIAA0879; NPP4; NPP-4 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
22875 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1.5 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
Q9Y6X5 | |
ENPP4 | |
Synthetic peptide corresponding to a region derived from internal residues of human ectonucleotide pyrophosphatase/phosphodiesterase 4 (putative) | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |