Learn More
Entecavir monohydrate, MedChemExpress
Product Code. 16496246
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
200 mg
Unit Size:
100mg
10mg
200mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Quantity Packungsgröße
16496246 100 mg 100mg
16478447 5 mg 5mg
16435778 10 mg 10mg
16421937 50 mg 50mg
16401096 200 mg 200mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Artikelnummer. 16496246 Lieferant Medchem Express Lieferanten-Nr. HY13623A
 Restricted Item
Verfügbar ab: 02-12-2026
Zum Warenkorb hinzufügen
Zum Warenkorb hinzufügen
missing translation for 'validMainframeMsg'
MedChemExpress Entecavir monohydrate (BMS200475 monohydrate; SQ34676 monohydrate) is a potent and selective inhibitor of HBV, with an EC50 of 3.75 nM in HepG2 cell.
Entecavir monohydrate (BMS200475 monohydrate
Spezifikation
Chemical Name or Material Entecavir monohydrate
CAS 209216-23-9
Color Off-White
Purity Grade Notes Research
Molecular Formula C12H17N5O4
Quantity 100 mg
Synonym BMS200475 monohydrateSQ34676 monohydrate
Solubility Information DMSO : ≥ 50 mg/mL (169.33 mM) ∣H2O : 2.8 mg/mL (9.48 mM; Need ultrasonic and warming)
SMILES O=C1NC(N)=NC2=C1N=CN2[C@@H]3C([C@H](CO)[C@@H](O)C3)=C.O
Molecular Weight (g/mol) 295.29
Formula Weight 295.29
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-anti-virus