missing translation for 'onlineSavingsMsg'
Learn More

ERAB Antibody, Novus Biologicals™

Artikelnummer. 18527123 Alle ansehen Bio Techne Produkte
Change view
Click to view available options
Menge:
0.025 mg
Packungsgröße:
25 Mikrogramm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge unitSize
18527123 0.025 mg 25 Mikrogramm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 18527123 Lieferant Novus Biologicals Lieferanten-Nr. NB100754600.025mg

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Chicken Polyclonal Antibody

ERAB Polyclonal specifically detects ERAB in Human, Mouse, Rat samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY

Spezifikation

Antigen ERAB
Anwendungen Western Blot
Klassifikation Polyclonal
Konjugat Unconjugated
Zusammensetzung PBS with No Preservative
Gen-Zugriffsnummer Q16836
Gen-Alias 17-beta-HSD 10, 17-beta-hydroxysteroid dehydrogenase 10, 17b-HSD10, 3-hydroxy-2-methylbutyryl-CoA dehydrogenase, ABAD, amyloid-beta peptide binding alcohol dehydrogenase, CAMR, DUPXp11.22, EC 1.1.1.178, EC 1.1.1.35, Endoplasmic reticulum-associated amyloid beta-peptide-binding protein, ERAB3-hydroxyacyl-CoA dehydrogenase type-2, HADH2AB-binding alcohol dehydrogenase, hydroxyacyl-Coenzyme A dehydrogenase, type II, hydroxyacyl-Coenzyme Adehydrogenase, type II, hydroxysteroid (17-beta) dehydrogenase 10, mental retardation, X-linked, syndromic 10, MHBD, Mitochondrial ribonuclease P protein 2,3-hydroxyacyl-CoA dehydrogenase type II, Mitochondrial RNase P protein 2, MRPP2HCD2, MRX17, MRX31, MRXS10, SCHAD, SDR5C1, short chain dehydrogenase/reductase family 5C, member 1, short chain L-3-hydroxyacyl-CoA dehydrogenase type 2, short chain type dehydrogenase/reductase XH98G2, Short-chain type dehydrogenase/reductase XH98G2, Type II HADH, XH98G2
Gensymbole HSD17B10
Wirtsspezies Chicken
Immunogen The immunogen for this antibody was a sequence from 57a.a. to 314a.a. from ERAB(a 314a.a. long protein)
Reinigungsverfahren Affinity Purified
Menge 0.025 mg
Regulatorischer Status RUO
Forschungsgebiet Lipid and Metabolism
Primär oder sekundär Primary
Gen-ID (Entrez) 3028
Testspezifität ERAB
Zielspezies Human
Inhalt und Lagerung Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles.
Mehr anzeigen Weniger anzeigen

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.