missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ ERH (Human) Recombinant Protein
Human ERH full-length ORF ( AAH14301, 1 a.a. - 104 a.a.) recombinant protein with GST-tag at N-terminal.
Marke: Abnova™ H00002079-P01.10ug
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
- Sequence: MSHTILLVQPTKRPEGRTYADYESVNECMEGVCKMYEEHLKRMNPNSPSITYDISQLFDFIDDLADLSCLVYRADTQTYQPYNKDWIKEKIYVLLRRQAQQAGK
Spezifikation
AAH14301 | |
enhancer of rudimentary homolog (Drosophila) | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ERH | |
GST |
2079 | |
Wheat germ expression system | |
10 μg | |
DROER, FLJ27340 | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |