Learn More
Thermo Scientific™ ESI Probes
Product Code. 17852836 Shop All Thermo Scientific Products
Change view
Click to view available options
Includes:
Internal STD Probe, orange
Diameter (Metric) Inner:
0.5 mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Includes Diameter (Metric) Inner Unit Size
17852836 Internal STD Probe, orange 0.5 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17852836 Supplier Thermo Scientific™ Supplier No. 1301630
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here
missing translation for 'validMainframeMsg'
Choose a Thermo Scientific™ Autosampler Sample Probe to suit your matrix and improve elemental analysis results when using automated sample introduction.
Customized autosampler accessories ensure more efficient, reliable analysis for your autosampler and trace elemental analyzer. Choose the appropriate ESI probe from our comprehensive range of autosampler sample probes.
Choose an ESI autosampler sample probe from the following options: 
Standard Sample Probe
  • 0.5 mm ID—typically used with ICP-MS systems
  • 0.8 mm ID—typically used with ICP-OES systems
  • 1.0 mm ID—for special applications
Probes for use with ESI FAST and prepFAST Systems
  • 0.5 mm ID with black/black pump tubing attached—for transport of carrier solution
  • 0.5 mm ID with orange/green pump tubing attached—for transport of the internal standard
Specifications
For Use With (Equipment) ESI Autosamplers
Type Autosampler Sample Probe
Diameter (Metric) Inner 0.5 mm
Includes Internal STD Probe, orange