missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ESR2 Beta-6 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 55471-1-AP-150UL
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
The human ER-beta is a newly discovered estrogen receptor initially cloned and characterized from testis. The size and structure of ER-beta is very similar to ER-alpha with the ligand and DNA binding domains being highly conserved, while the amino terminus which serves as their transactivation domain has diverged significantly. Similar in function to ER-alpha ER-beta binds to estrogen with a high affinity and regulates estrogen specific gene activation through direct interaction with estrogen response elements (ERE′s).Spezifikation
ESR2 Beta-6 | |
Polyclonal | |
Unconjugated | |
ESR2 | |
ER BETA, Erb, ESR BETA, ESR2, ESRB, ESTRB, estrogen receptor 2 (ER beta), Estrogen receptor beta, NR3A2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
13983, 2100 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.34 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O08537, Q92731 | |
ESR2 | |
Peptide | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |