missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethanol, ≥99.8 % (GC), Honeywell™
Click to view available options
Menge:
4 x 2.5 l
Spezifikation
Chemischer Name oder Material | Ethanol |
CAS | 64-17-5 |
Summenformel | C2H6O |
Strukturformel | CH3CH2OH |
MDL-Nummer | MFCD00003568 |
UN-Nummer | UN1170 |
Beilstein | 1718733 |
Synonym | Ethylalkohol, Alkohol, Getreidealkohol, Ethylhydroxid, Ethylhydrat, ethyl hydrate, algrain, alcohol, anhydrol, tecsol |
InChI-Schlüssel | LFQSCWFLJHTTHZ-UHFFFAOYSA-N |
SMILES | CCO |
Mehr anzeigen |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'