missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethanol (Taxed), Absolute, Reag. ISO, Reag. Ph. Eur., liquid, ≥99.8% (GC), for domestic sale in Germany, France, Ireland and United Kingdom, Honeywell Riedel-de Haën™
Click to view available options
Quantity:
500 mL
1 L
2.5 L
25 L
4 x 2.5 L
160 kg
Unit Size:
1 L
160kg
2.5 L
25 L
500mL
Pack of 4
Specifications
Specifications
| Density | 0.789g/mL at 25°C |
| Quantity | 2.5 L |
| Explosion Limit | 19% (60°F) |
| Flash Point | 14°C (57.2°F) |
| Ignition Point | 683°F |
| Physical Form | Liquid |
| Linear Formula | CH3CH2OH |
| Vapor Pressure | 44.6mmHg (20°C) |
| UN Number | 1170 |
| Beilstein | 1718733 |
| Show More |
Safety and Handling
Product Identifier
- Ethanol
- Danger
- Flammable liquid Category 2
- Serious eye damage/eye irritation Category 2
- H225-Highly flammable liquid and vapour.
- H319-Causes serious eye irritation.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction