Learn More
Ethanol D6 99.00% D, Eurisotop
Product Code. 17287436
Encompass
Change view
Click to view available options
Quantity:
5 mL
2 x 1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
17287436 5 mL
15941320 2 x 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: This product is subject to excise duty on alcohol in accordance with European Directive 92/83/EEC, all customers must have an authorization before delivery.
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 17287436 Supplier Eurisotop Supplier No. D114FD
 Restricted Item
Verfügbar ab: 11-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Regulatory: This product is subject to excise duty on alcohol in accordance with European Directive 92/83/EEC, all customers must have an authorization before delivery.
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Synonym: Ethanol-d6, Ethyl alcohol-d6, Ethyl-d5 alcohol-d, Hexadeuteroethanol
Ethanol D6
  • Isotopic Enrichment: 99.00% D
CoA and MSDS available at Eurisotop.com
Chemical Identifiers
CAS 1516-08-1
Molecular Formula C2H6O
Molecular Weight (g/mol) 52.11
InChI Key LFQSCWFLJHTTHZ-LIDOUZCJSA-N
Synonym Ethanol-d6, Ethyl alcohol-d6, Ethyl-d5 alcohol-d, Hexadeuteroethanol
IUPAC Name (2H5)ethan-1-(2H)ol
SMILES [2H]OC([2H])([2H])C([2H])([2H])[2H]
Specifications
Color Colorless
Deuterium (D) 99.00% D
Quantity 5 mL
Melting Point -114.1°C
Boiling Point 78.5°C
Grade Research
Chemical Name or Material Ethanol D6