missing translation for 'onlineSavingsMsg'
Learn More

Ethyl-1,2,3-thiadiazol-4-carboxylat, 97 %, Thermo Scientific™

Catalog Number p-7024727
Click to view available options
Menge:
1 g
5 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
3989-36-4
C5H6N2O2S
158.175
MFCD00052100
FBUHTNOJXVICFM-UHFFFAOYSA-N
ethyl 1,2,3-thiadiazole-4-carboxylate, acmc-209wkn, maybridge1_007257, 4-thiadiazolecarboxylic acid ethyl ester, ethyl 1,2,3-thiadiazol-4-ylcarboxylate, 1,2,3-thiadiazole-4-carboxylicacid, ethyl ester
254337
Ethylthiadiazol-4-Carboxylat
CCOC(=O)C1=CSN=N1
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
3989-36-4
C5H6N2O2S
158.175
MFCD00052100
FBUHTNOJXVICFM-UHFFFAOYSA-N
ethyl 1,2,3-thiadiazole-4-carboxylate, acmc-209wkn, maybridge1_007257, 4-thiadiazolecarboxylic acid ethyl ester, ethyl 1,2,3-thiadiazol-4-ylcarboxylate, 1,2,3-thiadiazole-4-carboxylicacid, ethyl ester
254337
Ethylthiadiazol-4-Carboxylat
CCOC(=O)C1=CSN=N1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material Ethyl 1,2,3-thiadiazole-4-carboxylate
Schmelzpunkt 83°C to 84°C
Menge 1 g
Formelmasse 158.18
Reinheit (%) 97%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'