missing translation for 'onlineSavingsMsg'
Learn More

1-Boc-DL-piperidin-3-carbonsäureethylester, 97 %, Thermo Scientific Chemicals

Catalog Number 15465457
Click to view available options
Menge:
1 g
5 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
130250-54-3
C13H23NO4
257.33
MFCD04116274
YCXCRFGBFZTUSU-UHFFFAOYSA-N
ethyl 1-boc-3-piperidinecarboxylate, 1-tert-butyl 3-ethyl piperidine-1,3-dicarboxylate, ethyl n-boc-piperidine-3-carboxylate, ethyl n-boc-3-piperidinecarboxylate, ethyl1-boc-3-piperidinecarboxylate, 1-boc-piperidine-3-carboxylic acid ethyl ester, ethyl-n-boc-piperidine-3-carboxylate, 1-tert-butyl 3-ethyl tetrahydro-1,3 2h-pyridinedicarboxylate, 1,3-piperidinedicarboxylic acid, 1-1,1-dimethylethyl 3-ethyl ester, piperidine-1,3-dicarboxylic acid 1-tert-butyl 3-ethyl ester
357727
1-O-Tert-Butyl 3-O-Ethylpiperidin-1,3-Dicarboxylat
CCOC(=O)C1CCCN(C1)C(=O)OC(C)(C)C
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
130250-54-3
C13H23NO4
257.33
MFCD04116274
YCXCRFGBFZTUSU-UHFFFAOYSA-N
ethyl 1-boc-3-piperidinecarboxylate, 1-tert-butyl 3-ethyl piperidine-1,3-dicarboxylate, ethyl n-boc-piperidine-3-carboxylate, ethyl n-boc-3-piperidinecarboxylate, ethyl1-boc-3-piperidinecarboxylate, 1-boc-piperidine-3-carboxylic acid ethyl ester, ethyl-n-boc-piperidine-3-carboxylate, 1-tert-butyl 3-ethyl tetrahydro-1,3 2h-pyridinedicarboxylate, 1,3-piperidinedicarboxylic acid, 1-1,1-dimethylethyl 3-ethyl ester, piperidine-1,3-dicarboxylic acid 1-tert-butyl 3-ethyl ester
357727
1-O-Tert-Butyl 3-O-Ethylpiperidin-1,3-Dicarboxylat
CCOC(=O)C1CCCN(C1)C(=O)OC(C)(C)C

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material Ethyl 1-Boc-DL-nipecotate
Schmelzpunkt 31°C to 35°C
Siedepunkt 95°C (0.5 mmHg)
Menge 1 g
Empfindlichkeit Light and moisture sensitive
Formelmasse 257.33
Reinheit (%) 97%

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'