missing translation for 'onlineSavingsMsg'
Learn More

Ethyl2-(1-Fmoc-3-indolyl)-4-methylthiazol-5-carboxylat, 97 %, Thermo Scientific™

Catalog Number p-7039203
Click to view available options
Menge:
1 g
5 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
1858255-80-7
C30H24N2O4S
508.592
MFCD22988949
RRACILVOHVVHHF-UHFFFAOYSA-N
9h-fluoren-9-ylmethyl 3-5-ethoxycarbonyl-4-methyl-1,3-thiazol-2-yl indole-1-carboxylate, ethyl 2-1-fmoc-3-indolyl-4-methylthiazole-5-carboxylate, ethyl 2-1-9h-fluoren-9-yl methoxy carbonyl-1h-indol-3-yl-4-methylthiazole-5-carboxylate
71741368
Ethyl2-[1-(9H-fluoren-9-ylmethoxycarbonyl)indol-3-yl]-4-methyl-1,3-thiazol-5-carboxylat
CCOC(=O)C1=C(N=C(S1)C2=CN(C3=CC=CC=C32)C(=O)OCC4C5=CC=CC=C5C6=CC=CC=C46)C
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
1858255-80-7
C30H24N2O4S
508.592
MFCD22988949
RRACILVOHVVHHF-UHFFFAOYSA-N
9h-fluoren-9-ylmethyl 3-5-ethoxycarbonyl-4-methyl-1,3-thiazol-2-yl indole-1-carboxylate, ethyl 2-1-fmoc-3-indolyl-4-methylthiazole-5-carboxylate, ethyl 2-1-9h-fluoren-9-yl methoxy carbonyl-1h-indol-3-yl-4-methylthiazole-5-carboxylate
71741368
Ethyl2-[1-(9H-fluoren-9-ylmethoxycarbonyl)indol-3-yl]-4-methyl-1,3-thiazol-5-carboxylat
CCOC(=O)C1=C(N=C(S1)C2=CN(C3=CC=CC=C32)C(=O)OCC4C5=CC=CC=C5C6=CC=CC=C46)C

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material Ethyl 2-(1-Fmoc-3-indolyl)-4-methylthiazole-5-carboxylate
Menge 1 g
Formelmasse 510.61
Reinheit (%) 97%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'