missing translation for 'onlineSavingsMsg'
Learn More

Ethyl-2-Amino-4-(2-Furyl)thiophen-3-Carboxylat, 97 %, Thermo Scientific™

Catalog Number p-7039478
Click to view available options
Menge:
2 g
10 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
256506-99-7
C11H11NO3S
237.27
MFCD00126394
JMLBJOYPNPOBKV-UHFFFAOYSA-N
ethyl 2-amino-4-2-furyl thiophene-3-carboxylate, ethyl 2-amino-4-furan-2-yl thiophene-3-carboxylate, 3-thiophenecarboxylicacid, 2-amino-4-2-furanyl-, ethyl ester, ethyl 2-amino-4-2-furyl-3-thiophenecarboxylate, ethyl 2-azanyl-4-furan-2-yl thiophene-3-carboxylate, 2-amino-4-2-furanyl-3-thiophenecarboxylic acid ethyl ester, 2-amino-4-furan-2-yl-thiophene-3-carboxylic acid ethyl ester
736628
ethyl 2-amino-4-(furan-2-yl)thiophene-3-carboxylate
CCOC(=O)C1=C(N)SC=C1C1=CC=CO1
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
256506-99-7
C11H11NO3S
237.27
MFCD00126394
JMLBJOYPNPOBKV-UHFFFAOYSA-N
ethyl 2-amino-4-2-furyl thiophene-3-carboxylate, ethyl 2-amino-4-furan-2-yl thiophene-3-carboxylate, 3-thiophenecarboxylicacid, 2-amino-4-2-furanyl-, ethyl ester, ethyl 2-amino-4-2-furyl-3-thiophenecarboxylate, ethyl 2-azanyl-4-furan-2-yl thiophene-3-carboxylate, 2-amino-4-2-furanyl-3-thiophenecarboxylic acid ethyl ester, 2-amino-4-furan-2-yl-thiophene-3-carboxylic acid ethyl ester
736628
ethyl 2-amino-4-(furan-2-yl)thiophene-3-carboxylate
CCOC(=O)C1=C(N)SC=C1C1=CC=CO1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material Ethyl 2-amino-4-(2-furyl)thiophene-3-carboxylate
Menge 2 g
Formelmasse 237.27
Reinheit (%) 97%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'