missing translation for 'onlineSavingsMsg'
Learn More

Ethyl-Amino-2-4,5-Dimethoxybenzoat, 98 %

Catalog Number p-7043743
Click to view available options
Menge:
1 g
5 g
25 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
20323-74-4
C11H15NO4
225.244
MFCD00017323
SMICMEHDDWELMR-UHFFFAOYSA-N
ethyl 6-amino-3,4-dimethoxybenzoate, benzoic acid, 2-amino-4,5-dimethoxy-, ethyl ester, 6-amino-3,4-dimethoxybenzoic acid ethyl ester, 2-amino-4,5-dimethoxybenzoic acid ethyl ester, ethyl 4,5-dimethoxyanthranilate, ethyl2-amino-4,5-dimethoxybenzoate, acmc-20a83g
88488
Ethyl2-amino-4,5-dimethoxybenzoat
CCOC(=O)C1=CC(=C(C=C1N)OC)OC
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
20323-74-4
C11H15NO4
225.244
MFCD00017323
SMICMEHDDWELMR-UHFFFAOYSA-N
ethyl 6-amino-3,4-dimethoxybenzoate, benzoic acid, 2-amino-4,5-dimethoxy-, ethyl ester, 6-amino-3,4-dimethoxybenzoic acid ethyl ester, 2-amino-4,5-dimethoxybenzoic acid ethyl ester, ethyl 4,5-dimethoxyanthranilate, ethyl2-amino-4,5-dimethoxybenzoate, acmc-20a83g
88488
Ethyl2-amino-4,5-dimethoxybenzoat
CCOC(=O)C1=CC(=C(C=C1N)OC)OC

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material Ethyl 2-amino-4,5-dimethoxybenzoate
Schmelzpunkt 86°C to 88°C
Menge 1 g
Beilstein 2646663
Formelmasse 225.24
Reinheit (%) 98%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'