missing translation for 'onlineSavingsMsg'
Learn More

Ethyl-2-(Ethoxycarbonylmethoxy)benzoat, 97 %, Thermo Scientific™

Catalog Number p-7046197
Click to view available options
Menge:
10 g
50 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
56424-77-2
C13H16O5
252.266
MFCD00151820
NIUVYKIVCCMXLR-UHFFFAOYSA-N
ethyl 2-2-ethoxy-2-oxoethoxy benzoate, ethyl 2-ethoxycarbonylmethoxy benzoate, ethyl o-carboethoxymethylsalicylate, ethyl 2-2-ethoxy-2-oxo-ethoxy benzoate, ethyl 2-2-ethoxycarbonyl phenoxy acetate, 2-2-ethoxy-2-oxoethoxy benzoic acid ethyl ester, ethyl 2-2-ethoxy-2-oxidanylidene-ethoxy benzoate, benzoic acid,2-2-ethoxy-2-oxoethoxy-, ethyl ester
347810
Ethyl2-(2-ethoxy-2-oxoethoxy)benzoat
CCOC(=O)COC1=CC=CC=C1C(=O)OCC
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
56424-77-2
C13H16O5
252.266
MFCD00151820
NIUVYKIVCCMXLR-UHFFFAOYSA-N
ethyl 2-2-ethoxy-2-oxoethoxy benzoate, ethyl 2-ethoxycarbonylmethoxy benzoate, ethyl o-carboethoxymethylsalicylate, ethyl 2-2-ethoxy-2-oxo-ethoxy benzoate, ethyl 2-2-ethoxycarbonyl phenoxy acetate, 2-2-ethoxy-2-oxoethoxy benzoic acid ethyl ester, ethyl 2-2-ethoxy-2-oxidanylidene-ethoxy benzoate, benzoic acid,2-2-ethoxy-2-oxoethoxy-, ethyl ester
347810
Ethyl2-(2-ethoxy-2-oxoethoxy)benzoat
CCOC(=O)COC1=CC=CC=C1C(=O)OCC

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material Ethyl 2-(ethoxycarbonylmethoxy)benzoate
Siedepunkt 180°C (4 mmHg)
Brechungsindex 1.514
Menge 10 g
Beilstein 2659411
Formelmasse 252.27
Reinheit (%) 97%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'