missing translation for 'onlineSavingsMsg'
Learn More

Ethyl3-(2-trimethylacetamido-3-pyridyl)acrylat, Thermo Scientific™

Catalog Number p-7033016
Click to view available options
Menge:
1 g
5 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
882029-13-2
C15H20N2O3
276.34
MFCD07781145
FRJWOJJLXVTBCQ-UHFFFAOYSA-N
ethyl 3-2-trimethylacetamido-3-pyridyl acrylate, ethyl 2e-3-2-2,2-dimethylpropanamido pyridin-3-yl prop-2-enoate, ethyl 3-2-pivalamidopyridin-3-yl acrylate, ethyl 3-2-pivaloylamino-3-pyridyl acrylate, 2-pivaloylamino pyridine-3-acrylic acid ethyl ester, ethyl 3-2-pivaloylamino pyridin-3-yl acrylate, 3-2-trimethylacetamido-3-pyridyl acrylic acid ethyl ester, ethyl 2e-3-2-2,2-dimethylpropanoylamino 3-pyridyl prop-2-enoate, 2-propenoic acid,3-2-2,2-dimethyl-1-oxopropyl amino-3-pyridinyl-,ethyl ester
44721296
ethyl 3-[2-(2,2-dimethylpropanamido)pyridin-3-yl]prop-2-enoate
CCOC(=O)C=CC1=C(NC(=O)C(C)(C)C)N=CC=C1
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
882029-13-2
C15H20N2O3
276.34
MFCD07781145
FRJWOJJLXVTBCQ-UHFFFAOYSA-N
ethyl 3-2-trimethylacetamido-3-pyridyl acrylate, ethyl 2e-3-2-2,2-dimethylpropanamido pyridin-3-yl prop-2-enoate, ethyl 3-2-pivalamidopyridin-3-yl acrylate, ethyl 3-2-pivaloylamino-3-pyridyl acrylate, 2-pivaloylamino pyridine-3-acrylic acid ethyl ester, ethyl 3-2-pivaloylamino pyridin-3-yl acrylate, 3-2-trimethylacetamido-3-pyridyl acrylic acid ethyl ester, ethyl 2e-3-2-2,2-dimethylpropanoylamino 3-pyridyl prop-2-enoate, 2-propenoic acid,3-2-2,2-dimethyl-1-oxopropyl amino-3-pyridinyl-,ethyl ester
44721296
ethyl 3-[2-(2,2-dimethylpropanamido)pyridin-3-yl]prop-2-enoate
CCOC(=O)C=CC1=C(NC(=O)C(C)(C)C)N=CC=C1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material Ethyl 3-(2-trimethylacetamido-3-pyridyl)acrylate
Menge 1 g
Formelmasse 276.34
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'