Learn More
Alfa Aesar™ Ethyl4-Acetylbutyrat, 97 %
Marke: Alfa Aesar™ L17848.14
Weitere Details : CAS : 13984-57-1 Netto-Gewicht : 0.02500kg
Menge | 25g |
---|
Chemische Identifikatoren
13984-57-1 | |
158.197 | |
MGPSIDGTLFKDEY-UHFFFAOYSA-N | |
84130 | |
CCOC(=O)CCCC(=O)C |
C8H14O3 | |
MFCD00009213 | |
ethyl 4-acetylbutyrate, hexanoic acid, 5-oxo-, ethyl ester, 5-oxohexanoic acid ethyl ester, ethyl 4-acetylbutanoate, ethyl 4-acetyl-butanoate, ethyl 4-acetyl-butyrate, acmc-1c2gr, 5-oxo-hexanoic acid ethyl ester, mgpsidgtlfkdey-uhfffaoysa | |
Ethyl-5-oxohexanoat |
Spezifikation
Ethyl 4-acetylbutyrate | |
69°C (156°F) | |
65°C (0.76mmHg) | |
13984-57-1 | |
MFCD00009213 | |
ethyl 4-acetylbutyrate, hexanoic acid, 5-oxo-, ethyl ester, 5-oxohexanoic acid ethyl ester, ethyl 4-acetylbutanoate, ethyl 4-acetyl-butanoate, ethyl 4-acetyl-butyrate, acmc-1c2gr, 5-oxo-hexanoic acid ethyl ester, mgpsidgtlfkdey-uhfffaoysa | |
CCOC(=O)CCCC(=O)C | |
158.197 | |
158.2 |
0.989 | |
1.4275 | |
25g | |
C8H14O3 | |
1762625 | |
MGPSIDGTLFKDEY-UHFFFAOYSA-N | |
Ethyl-5-oxohexanoat | |
84130 | |
97% |