missing translation for 'onlineSavingsMsg'
Learn More

Ethyl-4-methyl-2-(1H-pyrazol-1-yl)-thiazol-5-carboxylat, 97 %, Thermo Scientific™

Catalog Number p-7039147
Click to view available options
Menge:
1 g
5 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
1240328-26-0
C10H11N3O2S
237.277
MFCD22988945
QIEQTJFJYQVLIU-UHFFFAOYSA-N
ethyl 4-methyl-2-pyrazol-1-yl-1,3-thiazole-5-carboxylate, ethyl 4-methyl-2-1h-pyrazol-1-yl thiazole-5-carboxylate
98000048
Ethyl 4-Methyl-2-Pyrazol-1-yl-1,3-Thiazol-5-Carboxylat
CCOC(=O)C1=C(N=C(S1)N2C=CC=N2)C
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
1240328-26-0
C10H11N3O2S
237.277
MFCD22988945
QIEQTJFJYQVLIU-UHFFFAOYSA-N
ethyl 4-methyl-2-pyrazol-1-yl-1,3-thiazole-5-carboxylate, ethyl 4-methyl-2-1h-pyrazol-1-yl thiazole-5-carboxylate
98000048
Ethyl 4-Methyl-2-Pyrazol-1-yl-1,3-Thiazol-5-Carboxylat
CCOC(=O)C1=C(N=C(S1)N2C=CC=N2)C

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material Ethyl 4-methyl-2-(1H-pyrazol-1-yl)thiazole-5-carboxylate
Menge 1 g
Formelmasse 237.28
Reinheit (%) 97%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'