Learn More
Ethyl-4-methyl-2-(2-pyridyl)-thiazol-5-carboxylat, 97 %, Thermo Scientific™
Catalog Number p-7036534
Click to view available options
Menge:
250 mg
1 g
5 g
383146-88-1
C12H12N2O2S
248.30
MFCD01936157
UGCVLLXXBVTJPK-UHFFFAOYSA-N
ethyl 4-methyl-2-pyridin-2-yl thiazole-5-carboxylate, ethyl 4-methyl-2-pyridin-2-yl-1,3-thiazole-5-carboxylate, ethyl 4-methyl-2-2-pyridinyl-1,3-thiazole-5-carboxylate, ethyl 4-methyl-2-pyridin-2-ylthiazole-5-carboxylate, ethyl 4-methyl-2-2-pyridyl-1,3-thiazole-5-carboxylate, ethyl 4-methyl-2-2-pyridyl thiazole-5-carboxylate, ethyl4-methyl-2-pyridin-2-yl thiazole-5-carboxylate
2763623
Ethyl 4-Methyl-2-Pyridin-2-yl-1,3-Thiazol-5-Carboxylat
CCOC(=O)C1=C(C)N=C(S1)C1=CC=CC=N1
383146-88-1
C12H12N2O2S
248.30
MFCD01936157
UGCVLLXXBVTJPK-UHFFFAOYSA-N
ethyl 4-methyl-2-pyridin-2-yl thiazole-5-carboxylate, ethyl 4-methyl-2-pyridin-2-yl-1,3-thiazole-5-carboxylate, ethyl 4-methyl-2-2-pyridinyl-1,3-thiazole-5-carboxylate, ethyl 4-methyl-2-pyridin-2-ylthiazole-5-carboxylate, ethyl 4-methyl-2-2-pyridyl-1,3-thiazole-5-carboxylate, ethyl 4-methyl-2-2-pyridyl thiazole-5-carboxylate, ethyl4-methyl-2-pyridin-2-yl thiazole-5-carboxylate
2763623
Ethyl 4-Methyl-2-Pyridin-2-yl-1,3-Thiazol-5-Carboxylat
CCOC(=O)C1=C(C)N=C(S1)C1=CC=CC=N1
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemischer Name oder Material Ethyl 4-methyl-2-(2-pyridyl)thiazole-5-carboxylate
Menge 250 mg
Formelmasse 248.3
Reinheit (%) 97%