missing translation for 'onlineSavingsMsg'
Learn More

Ethyl-5-Phenoxycarbonylamino-1,2,3-Thiadiazol-4-Carboxylat, 98+ %, Thermo Scientific Chemicals

Catalog Number p-7028440
Click to view available options
Menge:
250 mg
1 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
2037-81-2
C12H11N3O4S
293.30
MFCD00100230
FCPHNPRVMWSGSK-UHFFFAOYSA-N
ethyl 5-phenoxycarbonylamino-1,2,3-thiadiazole-4-carboxylate, ethyl 5-phenoxycarbonyl amino-1,2,3-thiadiazole-4-carboxylate, 5-n-phenoxycarbonylamino-1,2,3-thiadiazole-4-carboxylic acid ethyl ester, tos-bb-0370, ethyl 5-phenoxycarbonylamino thiadiazole-4-carboxylate, ethyl 5-phenoxycarbonylamino-1,2,3-thiadiazole-4-carboxylate, 98+%
6917126
Ethyl5-(phenoxycarbonylamino)thiadiazol-4-carboxylat
CCOC(=O)C1=C(NC(=O)OC2=CC=CC=C2)SN=N1
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
2037-81-2
C12H11N3O4S
293.30
MFCD00100230
FCPHNPRVMWSGSK-UHFFFAOYSA-N
ethyl 5-phenoxycarbonylamino-1,2,3-thiadiazole-4-carboxylate, ethyl 5-phenoxycarbonyl amino-1,2,3-thiadiazole-4-carboxylate, 5-n-phenoxycarbonylamino-1,2,3-thiadiazole-4-carboxylic acid ethyl ester, tos-bb-0370, ethyl 5-phenoxycarbonylamino thiadiazole-4-carboxylate, ethyl 5-phenoxycarbonylamino-1,2,3-thiadiazole-4-carboxylate, 98+%
6917126
Ethyl5-(phenoxycarbonylamino)thiadiazol-4-carboxylat
CCOC(=O)C1=C(NC(=O)OC2=CC=CC=C2)SN=N1

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material Ethyl 5-phenoxycarbonylamino-1,2,3-thiadiazole-4-carboxylate
Schmelzpunkt 153°C to 155°C
Menge 250 mg
Formelmasse 293.3
Reinheit (%) ≥98%

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'