missing translation for 'onlineSavingsMsg'
Learn More

Ethyl alpha,alpha-Difluorphenylacetat, 99%, Thermo Scientific™

Catalog Number p-7028635
Click to view available options
Menge:
1 g
5 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
2248-46-6
C10H10F2O2
200.185
MFCD08275296
KCMSDCHUELUJPX-UHFFFAOYSA-N
ethyl2,2-difluoro-2-phenylacetate, ethyl difluoro phenyl acetate, alpha,alpha-difluoro phenylacetic acid ethyl ester, difluoro-phenyl-acetic acid ethyl ester, ethyl 2,2-difluoro-2-phenyl-acetate, acmc-1cmi4, difluorophenylacetic acid ethyl ester, ethyl 2,2-difluoro-2-phenylethanoate, ethyl alpha,alpha-difluorophenylacetate, difluoro phenyl acetic acid ethyl ester
10631860
Ethyl-2,2-difluor-2-phenylacetat
CCOC(=O)C(C1=CC=CC=C1)(F)F
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
2248-46-6
C10H10F2O2
200.185
MFCD08275296
KCMSDCHUELUJPX-UHFFFAOYSA-N
ethyl2,2-difluoro-2-phenylacetate, ethyl difluoro phenyl acetate, alpha,alpha-difluoro phenylacetic acid ethyl ester, difluoro-phenyl-acetic acid ethyl ester, ethyl 2,2-difluoro-2-phenyl-acetate, acmc-1cmi4, difluorophenylacetic acid ethyl ester, ethyl 2,2-difluoro-2-phenylethanoate, ethyl alpha,alpha-difluorophenylacetate, difluoro phenyl acetic acid ethyl ester
10631860
Ethyl-2,2-difluor-2-phenylacetat
CCOC(=O)C(C1=CC=CC=C1)(F)F

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material Ethyl alpha,alpha-difluorophenylacetate
Brechungsindex 1.46
Menge 1 g
Formelmasse 200.19
Reinheit (%) 99%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'