missing translation for 'onlineSavingsMsg'
Learn More

Ethylchlorfluoracetat, 98%, Thermo Scientific™

Catalog Number p-7021738
Click to view available options
Menge:
5 g
25 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
401-56-9
C4H6ClFO2
140.538
MFCD00039333
WUHVJSONZHSDFC-UHFFFAOYSA-N
ethyl chlorofluoroacetate, acetic acid, chlorofluoro-, ethyl ester, chlorofluoroacetic acid ethyl ester, ethyl chloro fluoro acetate, ethyl fluorochloroacetate, acetic acid, 2-chloro-2-fluoro-, ethyl ester, acmc-1csjg, ksc492m6l, ethyl 2-chloro-2-fluoro-acetate
101668
Ethyl-2-chlor-2-fluoracetat
CCOC(=O)C(F)Cl
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
401-56-9
C4H6ClFO2
140.538
MFCD00039333
WUHVJSONZHSDFC-UHFFFAOYSA-N
ethyl chlorofluoroacetate, acetic acid, chlorofluoro-, ethyl ester, chlorofluoroacetic acid ethyl ester, ethyl chloro fluoro acetate, ethyl fluorochloroacetate, acetic acid, 2-chloro-2-fluoro-, ethyl ester, acmc-1csjg, ksc492m6l, ethyl 2-chloro-2-fluoro-acetate
101668
Ethyl-2-chlor-2-fluoracetat
CCOC(=O)C(F)Cl

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material Ethyl chlorofluoroacetate
Dichte 1.225
Siedepunkt 128°C to 130°C
Flammpunkt 44°C (111°F)
Geruch Pungent
Brechungsindex 1.396
Menge 5 g
UN-Nummer UN2920
Beilstein 1748679
Formelmasse 140.54
Reinheit (%) 98%
Mehr anzeigen Weniger anzeigen
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'