missing translation for 'onlineSavingsMsg'
Learn More

Ethylen-(Diphenylmethylen)Glyzinat, 98 %, Thermo Scientific Chemicals

Catalog Number p-3757847
Click to view available options
Menge:
5 g
25 g
Verpackung:
Glasflasche
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
69555-14-2
C17H17NO2
267.32
MFCD00010590
QUGJYNGNUBHTNS-UHFFFAOYSA-N
ethyl n-diphenylmethylene glycinate, n-diphenylmethylene glycine ethyl ester, ethyl 2-diphenylmethylene amino acetate, ethyl 2-diphenylmethyleneamino acetate, diphenylmethylene-glycine ethyl ester, ethyl 2-benzhydrylideneamino acetate, ethyl 2-diphenylmethylidene amino acetate, diphenylmethylene glycine ethylester, ethyl diphenylmethylenamino acetate, diphenylmethylene glycine ethyl ester
319508
ethyl-2-(benzhydrylideneamino)acetat
CCOC(=O)CN=C(C1=CC=CC=C1)C2=CC=CC=C2
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
69555-14-2
C17H17NO2
267.32
MFCD00010590
QUGJYNGNUBHTNS-UHFFFAOYSA-N
ethyl n-diphenylmethylene glycinate, n-diphenylmethylene glycine ethyl ester, ethyl 2-diphenylmethylene amino acetate, ethyl 2-diphenylmethyleneamino acetate, diphenylmethylene-glycine ethyl ester, ethyl 2-benzhydrylideneamino acetate, ethyl 2-diphenylmethylidene amino acetate, diphenylmethylene glycine ethylester, ethyl diphenylmethylenamino acetate, diphenylmethylene glycine ethyl ester
319508
ethyl-2-(benzhydrylideneamino)acetat
CCOC(=O)CN=C(C1=CC=CC=C1)C2=CC=CC=C2

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material Ethyl N-(diphenylmethylene)glycinate
Schmelzpunkt 51°C to 55°C
Farbe Weiß bis gelb
Flammpunkt >110°C
Infrarotspektrum Echt
Prozentgehaltsbereich 97.5% min. (GC)
Verpackung Glasflasche
Strukturformel (C6H5)2C=NCH2COOCH2CH3
Menge 5 g
Formelmasse 267.32
Reinheit (%) 98%
Physikalische Form Kristallines Pulver oder Kristalle
Mehr anzeigen Weniger anzeigen

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'