Learn More
Invitrogen™ EVOS™ M3000 Attachable Mechanical Stage
Artikelnummer. 30178684
Ansicht ändern
Click to view available options
Menge:
1 Probentisch
Packungsgröße:
1 Stück
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge Packungsgröße
30178684 1 Probentisch 1 Stück
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Artikelnummer. 30178684 Lieferant Invitrogen™ Lieferanten-Nr. AMEP5011
Schwerpunkt: Regenerative Therapien
Verwenden Sie den Aktionscode 28027
Alle Werbeaktionen anzeigen
Verfügbar ab: 14-10-2026
Zum Warenkorb hinzufügen
Zum Warenkorb hinzufügen
missing translation for 'validMainframeMsg'
The EVOS M3000 Mechanical Stage is an optional addition to the universal, fixed stage provided with the EVOS M3000 Imaging System.
The EVOS M3000 Mechanical Stage is an optional addition to the universal, fixed stage provided with the EVOS M3000 Imaging System. This mechanical stage has X-Y axis controls and a vessel holder framework that accommodates our broad range of vessel holders.
The EVOS M3000 Mechanical Stage is an optional addition to the universal, fixed stage provided with the EVOS M3000 Imaging System. This mechanical stage has X-Y axis controls and a vessel holder framework that accommodates our broad range of vessel holders. Larger vessels, such as T-75 flasks or multi-well plates, fit directly onto the stage. Both vessel holders and large vessels are held securely in place by a stage clip during operation. The spring-loaded clip allows fast and easy vessel changes, such as during operations inside a hood or during processing of multiple samples.
TRUSTED_SUSTAINABILITY
Spezifikation
Typ Mechanical Stage
Zur Verwendung mit (Geräte) EVOS M3000 Imaging System
Menge 1 Probentisch
Nur für Forschungszwecke.