missing translation for 'onlineSavingsMsg'
Learn More
Learn More
F(ab')2-Goat anti-Human IgM Fc Secondary Antibody, Invitrogen™
F(ab')2-Goat Polyclonal Secondary Antibody
Marke: Invitrogen H15100
Weitere Details : Netto-Gewicht : 0.50000kg
Beschreibung
Human IgM Fc Polyclonal antibody specifically detects Human IgM Fc in Human samples. It is validated for Flow Cytometry.Spezifikation
Human IgM Fc, Fab′2-Goat anti-Human IgM Fc Secondary Antibody | |
Polyclonal | |
PBS with 0.1% sodium azide | |
purified | |
ASR | |
Human | |
Liquid |
Flow Cytometry | |
Unconjugated | |
Goat | |
1 mL | |
Secondary | |
4° C, store in dark | |
IgG |