Learn More
FAM198B Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA552406
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: PRTRRNLLLG TACAIYLGFL VSQVGRASLQ HGQAAEKGPH RSRDTAEPSF PEIPLDGTLA PPESQGNGST LQPNVVYITL RSKRSKPANI RGTVKPKRRK KHAVASAAPG QEALVGPSLQ PQEAAREADA VAPGYA Highest antigen sequence identity to the following orthologs: Mouse - 81%, Rat - 79%.
FAM198B Polyclonal antibody specifically detects FAM198B in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Spezifikation
FAM198B | |
Polyclonal | |
Unconjugated | |
FAM198B | |
1110032E23Rik; 2210419I08Rik; AD021; AD036; AV011458; C4orf18; Ened; Expressed in nerve and epithelium during development; FAM198B; family with sequence similarity 198 member B; family with sequence similarity 198, member B; gask1b; GaskbB; golgi associated kinase 1B; Golgi-associated kinase 1B; LOW QUALITY PROTEIN: protein FAM198B; protein ENED; protein ENED; protein FAM198B; protein FAM198B; UNQ2512/PRO6001 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
51313 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q6UWH4 | |
FAM198B | |
Recombinant protein corresponding to Human FAM198B | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only