Learn More
FAM55A Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA561890
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: YMTTRNREVS YLTDKENSLF HRSKVGVEMM KDRKHIDVTN CNKREKIEET CQVGMKPPVP Highest antigen sequence identity to the following orthologs: Mouse - 43%, Rat - 50%.
FAM55A Polyclonal antibody specifically detects FAM55A in Human samples. It is validated for Immunohistochemistry, Western Blot.Spezifikation
FAM55A | |
Polyclonal | |
Unconjugated | |
NXPE1 | |
FAM55A; family with sequence similarity 55, member A; neurexophilin and PC-esterase domain family member 1; neurexophilin and PC-esterase domain family, member 1; NXPE family member 1; NXPE1; Protein FAM55A | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
120400 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry, Western Blot | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8N323 | |
NXPE1 | |
Recombinant protein corresponding to Human FAM55A | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only