Learn More
FAM59A Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA559841
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: GNSEVNLHGC RDLGGDWAPF PHDILPYQDS GDSGSDYLFP EASEESAGIP GKSELPYEEL WLEEGKPSHQ PLTRSLSEKN RCDQFRGSVR SKCATSPLPI P Highest antigen sequence identity to the following orthologs: Mouse - 86%, Rat - 24%.
FAM59A Polyclonal antibody specifically detects FAM59A in Human samples. It is validated for Immunohistochemistry.Spezifikation
FAM59A | |
Polyclonal | |
Unconjugated | |
GAREM1 | |
C18orf11; C86169; Fam59a; family with sequence similarity 59, member A; GAREM; GAREM1; Gm944; GRB2 associated regulator of MAPK1 1; GRB2 associated regulator of MAPK1 subtype 1; GRB2 associated, regulator of MAPK1; Grb2-associated and regulator of Erk/MAPK; GRB2-associated and regulator of MAPK protein; GRB2-associated and regulator of MAPK protein 1; GRB2-associated and regulator of MAPK1; Kiaa4238; mKIAA4238; protein FAM59A | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
64762 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.8 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9H706 | |
GAREM1 | |
Recombinant protein corresponding to Human FAM59A | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only