missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FANCC Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 16973-1-AP-150UL
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
The Fanconi anemia complementation group (FANC) currently includes FANCA, FANCB, FANCC, FANCD1 (also called BRCA2), FANCD2, FANCE, FANCF, FANCG, FANCI, FANCJ (also called BRIP1), FANCL, FANCM and FANCN (also called PALB2). The previously defined group FANCH is the same as FANCA. Fanconi anemia is a genetically heterogeneous recessive disorder characterized by cytogenetic instability, hypersensitivity to DNA crosslinking agents, increased chromosomal breakage, and defective DNA repair. The members of the Fanconi anemia complementation group do not share sequence similarity; they are related by their assembly into a common nuclear protein complex. This gene encodes the protein for complementation group C.Spezifikation
FANCC | |
Polyclonal | |
Unconjugated | |
FANCC | |
FA3, FAC, FACC, FANCC, Fanconi anemia group C protein, Protein FACC | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
2176 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.21 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q00597 | |
FANCC | |
FANCC Fusion Protein Ag9423 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |