missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Fas Ligand/TNFSF6 Antibody, R&D Systems™
Mouse Monoclonal Antibody
N.V.
Spezifikation
Antigen | Fas Ligand/TNFSF6 |
---|---|
Klon | 154922 |
Konzentration | LYOPH |
Verdünnung | Western Blot 1 ug/mL |
Anwendungen | Western Blot |
Beschreibung
Fas Ligand/TNFSF6 Monoclonal specifically detects Fas Ligand/TNFSF6 in Human samples. It is validated for Western Blot.Spezifikation
Fas Ligand/TNFSF6 | |
LYOPH | |
Western Blot | |
Unconjugated | |
Mouse | |
Human | |
P48023 | |
356 | |
Chinese hamster ovary cell line CHO-derived recombinant human Fas Ligand Pro134-Leu281 Accession # P48023 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human Fas Ligand in direct ELISAs and Western blots. In Western blots, this antibody shows approximately 50% cross-reactivity with recombinant mouse (rm) Fas Ligand and less than 5% cross-reactivity with rhAPRIL, rhBAFF, rhEDA-A2, rhGITR Ligand, rhLIGHT, rhOX40 Ligand, rhTRAIL, rhTNF-alpha , rhTRANCE, rhTWEAK, or rhVEGI. |
154922 | |
Western Blot 1 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
apoptosis (APO-1) antigen ligand 1, Apoptosis antigen ligand, APT1LG1CD95L, APTL, CD178, CD178 antigen, CD95L, CD95-L, Fas antigen ligand, Fas ligand, Fas ligand (TNF superfamily, member 6), FASLCD95 ligand, FASLG, TNFSF6, TNFSF6FasL, tumor necrosis factor (ligand) superfamily, member 6, tumor necrosis factor ligand superfamily member 6 | |
FASLG | |
IgG1 | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only