missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-2',3'-dimethylazobenzol, Thermo Scientific Chemicals

Catalog Number p-7041875
Click to view available options
Menge:
100 g
500 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
97-56-3
C14H15N3
225.295
MFCD00007733
PFRYFZZSECNQOL-UHFFFAOYSA-N
C.I. 11160; Solvent Yellow 3
7340
2-Methyl-4-[(2-Methylphenyl)Diazenyl]Anilin
CC1=CC=CC=C1N=NC2=CC(=C(C=C2)N)C
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
97-56-3
C14H15N3
225.295
MFCD00007733
PFRYFZZSECNQOL-UHFFFAOYSA-N
C.I. 11160; Solvent Yellow 3
7340
2-Methyl-4-[(2-Methylphenyl)Diazenyl]Anilin
CC1=CC=CC=C1N=NC2=CC(=C(C=C2)N)C

C.I. 11160

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

C.I. 11160
TRUSTED_SUSTAINABILITY
Chemischer Name oder Material Fast Garnet GBC base
Schmelzpunkt 101°C to 102°C
Strukturformel CH3C6H4N=NC6H3(CH3)NH2
Beilstein 6506005
Merck Index 14,420
Formelmasse 225.29
Geruch Odorless
Farbe Braun-orange
Menge 100 g
Physikalische Form Pulver

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'