missing translation for 'onlineSavingsMsg'
Learn More

Fasudil hydrochloride, Tocris Bioscience™

Inhibitor of cyclic nucleotide dependent- and Rho-kinases

Marke:  Tocris (Bio-Techne) 054150

Weitere Details : Netto-Gewicht : 0.01000kg

Menge. 50mg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 13417703

  • 738.52 € / 50 Milligramm


Voraussichtlicher Liefertermin 01-08-2019
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

Cyclic nucleotide-dependent protein kinase inhibitor and Rho-associated kinase inhibitor (IC50 = 10.7 μM). Suppress MMP-2 expression and induce apoptosis in glioblastoma cells in vivo. Ca2+ antagonist and vasodilator. Also inhibits proliferation of vascular smooth muscle cells.
Spezifikation

Chemische Identifikatoren

105628-07-7
327.827
fasudil hydrochloride, fasudil hcl, eril, fasudil hydrochloride, at 877 hydrochloride, fasudil monohydrochloride, ha 1077 hydrochloride, unii-sq04n8s7br, 5-1,4-diazepan-1-yl sulfonyl isoquinoline hydrochloride, sq04n8s7br
5-(1,4-diazepan-1-ylsulfonyl)isoquinoline;hydrochloride
C14H18ClN3O2S
LFVPBERIVUNMGV-UHFFFAOYSA-N
163751
C1CNCCN(C1)S(=O)(=O)C2=CC=CC3=C2C=CN=C3.Cl

Spezifikation

Fasudil hydrochloride
105628-07-7
fasudil hydrochloride, fasudil hcl, eril, fasudil hydrochloride, at 877 hydrochloride, fasudil monohydrochloride, ha 1077 hydrochloride, unii-sq04n8s7br, 5-1,4-diazepan-1-yl sulfonyl isoquinoline hydrochloride, sq04n8s7br
C1CNCCN(C1)S(=O)(=O)C2=CC=CC3=C2C=CN=C3.Cl
327.827
327.83
50mg
C14H18ClN3O2S
LFVPBERIVUNMGV-UHFFFAOYSA-N
5-(1,4-diazepan-1-ylsulfonyl)isoquinoline;hydrochloride
163751
>98%
Videos